Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034407.1 | NP_001029579.1 | 7 - 8 | 1093 | 79 | |
GenBank mRNA | BC103405.1 | - | 7 - 8 | 1093 | 79 | |
BT030568.1 | - | 7 - 8 | 1077 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |