Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLGRKT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034426.2 | NP_001029598.1 | 5 - 6 | 569 | 77 | |
GenBank mRNA | BC102121.1 | - | 5 - 6 | 570 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |