Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034433.1 | NP_001029605.1 | 4 - 5 | 670 | 86 | |
GenBank mRNA | BC104546.1 | - | 4 - 5 | 670 | 86 | |
BT030507.1 | - | 4 - 5 | 682 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |