Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034479.1 | NP_001029651.1 | 2 - 3 | 235 | 81 | |
GenBank mRNA | DQ176047.1 | - | 2 - 3 | 235 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |