Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034497.2 | NP_001029669.1 | 4 - 5 | 904 | 66 | |
GenBank mRNA | BC102816.1 | - | 4 - 5 | 905 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |