Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034512.2 | NP_001029684.1 | 1 - 2 | 422 | 68 | |
GenBank mRNA | BC104569.1 | - | 1 - 2 | 427 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |