Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034515.1 | NP_001029687.1 | 2 - 3 | 1050 | 65 | |
GenBank mRNA | BC105147.1 | - | 2 - 3 | 1050 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |