Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034527.2 | NP_001029699.1 | 1 - 2 | 337 | 133 | |
GenBank mRNA | BC102464.1 | - | 1 - 2 | 338 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |