Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034535.1 | NP_001029707.1 | 1 - 2 | 113 | 80 | |
GenBank mRNA | BC103295.1 | - | 1 - 2 | 113 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |