Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034618.2 | NP_001029790.1 | 4 - 5 | 438 | 101 | |
GenBank mRNA | BC103352.1 | - | 4 - 5 | 439 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |