Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034618.2 | NP_001029790.1 | 6 - 7 | 624 | 73 | |
GenBank mRNA | BC103352.1 | - | 6 - 7 | 625 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |