Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034628.3 | NP_001029800.1 | 5 - 6 | 957 | 99 | |
NM_001320319.1 | NP_001307248.1 | 5 - 6 | 957 | 99 | ||
NR_135204.1 | 4 - 5 | 781 | 99 | |||
GenBank mRNA | BC102133.1 | - | 5 - 6 | 553 | 99 | |
DV882028.1 | - | 5 - 6 | 538 | 99 | ||
EH162161.1 | - | 4 - 5 | 403 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |