Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034662.2 | NP_001029834.1 | 4 - 5 | 401 | 85 | |
GenBank mRNA | BC102206.1 | - | 4 - 5 | 402 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |