Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034671.2 | NP_001029843.1 | 4 - 5 | 411 | 93 | |
GenBank mRNA | BC102423.1 | - | 4 - 5 | 412 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |