Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASNSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034680.2 | NP_001029852.1 | 1 - 2 | 164 | 102 | |
GenBank mRNA | BC103382.1 | - | 1 - 2 | 165 | 102 | |
BT026275.1 | - | 1 - 2 | 188 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |