Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034700.2 | NP_001029872.1 | 4 - 5 | 502 | 89 | |
GenBank mRNA | BC102735.1 | - | 4 - 5 | 504 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |