Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NICN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034767.2 | NP_001029939.1 | 5 - 6 | 648 | 64 | |
GenBank mRNA | BC105203.1 | - | 5 - 6 | 649 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |