Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIMAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034771.1 | NP_001029943.1 | 1 - 2 | 56 | 108 | |
GenBank mRNA | BC104533.1 | - | 1 - 2 | 56 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |