Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034795.1 | NP_001029967.1 | 5 - 6 | 816 | 74 | |
GenBank mRNA | BC102446.1 | - | 5 - 6 | 816 | 74 | |
BT020690.1 | - | 5 - 6 | 833 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |