Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034802.1 | NP_001029974.1 | 6 - 7 | 827 | 86 | |
GenBank mRNA | BC102183.1 | - | 6 - 7 | 827 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |