Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACYBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034809.2 | NP_001029981.1 | 2 - 3 | 343 | 108 | |
GenBank mRNA | BC102091.1 | - | 2 - 3 | 345 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |