Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACYBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034809.2 | NP_001029981.1 | 4 - 5 | 546 | 80 | |
GenBank mRNA | BC102091.1 | - | 4 - 5 | 548 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |