Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035045.1 | NP_001030217.1 | 1 - 2 | 951 | 78 | |
GenBank mRNA | BC105199.1 | - | 1 - 2 | 951 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |