Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035045.1 | NP_001030217.1 | 3 - 4 | 1232 | 107 | |
GenBank mRNA | BC105199.1 | - | 3 - 4 | 1232 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |