Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035094.2 | NP_001030266.1 | 4 - 5 | 532 | 88 | |
GenBank mRNA | BC102483.1 | - | 4 - 5 | 534 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |