Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035094.2 | NP_001030266.1 | 6 - 7 | 819 | 63 | |
GenBank mRNA | BC102483.1 | - | 6 - 7 | 821 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |