Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAO2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035110.1 | NP_001030282.1 | 1 - 2 | 154 | 64 | |
GenBank mRNA | BC102256.1 | - | 1 - 2 | 154 | 64 | |
BC151428.1 | - | 1 - 2 | 151 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |