Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035305.2 | NP_001030382.1 | 2 - 3 | 443 | 145 | |
GenBank mRNA | BC107540.1 | - | 2 - 3 | 444 | 145 | |
BT030747.1 | - | 2 - 3 | 336 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |