Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035307.2 | NP_001030384.1 | 2 - 3 | 226 | 66 | |
GenBank mRNA | BC102246.1 | - | 2 - 3 | 228 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |