Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205509.3 | NP_001192438.2 | 3 - 4 | 425 | 90 | |
GenBank mRNA | BC102673.1 | - | 3 - 4 | 422 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |