Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035381.2 | NP_001030458.1 | 2 - 3 | 268 | 71 | |
GenBank mRNA | BC103384.1 | - | 2 - 3 | 270 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |