Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035392.2 | NP_001030469.1 | 5 - 6 | 867 | 72 | |
GenBank mRNA | BC102669.1 | - | 5 - 6 | 868 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |