Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035431.2 | NP_001030508.1 | 2 - 3 | 233 | 80 | |
GenBank mRNA | BC102127.1 | - | 2 - 3 | 234 | 80 | |
BT030501.1 | - | 2 - 3 | 203 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |