Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035501.1 | NP_001030578.1 | 8 - 9 | 1074 | 99 | |
GenBank mRNA | BC103272.1 | - | 8 - 9 | 1074 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |