Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037319.1 | NP_001032396.1 | 4 - 5 | 1422 | 118 | |
GenBank mRNA | BC104598.2 | - | 4 - 5 | 1422 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |