Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUV39H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037479.2 | NP_001032556.1 | 2 - 3 | 196 | 125 | |
GenBank mRNA | BC108111.1 | - | 2 - 3 | 197 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |