Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037481.1 | NP_001032558.1 | 4 - 5 | 372 | 101 | |
NM_001113256.1 | NP_001106727.1 | 5 - 6 | 475 | 101 | ||
GenBank mRNA | BC108190.1 | - | 4 - 5 | 372 | 101 | |
BI849286.1 | - | 5 - 6 | 461 | 101 | ||
CN433599.1 | - | 5 - 6 | 395 | 101 | ||
DY212243.1 | - | 5 - 6 | 549 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |