Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037486.2 | NP_001032563.1 | 3 - 4 | 320 | 72 | |
NM_001164221.1 | NP_001157693.1 | 4 - 5 | 431 | 72 | ||
GenBank mRNA | BC108197.1 | - | 3 - 4 | 320 | 72 | |
EV663472.1 | - | 4 - 5 | 413 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |