Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038540.1 | NP_001033629.1 | 3 - 4 | 317 | 114 | |
GenBank mRNA | BC109756.1 | - | 3 - 4 | 317 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |