Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040472.2 | NP_001035562.2 | 1 - 2 | 127 | 82 | |
GenBank mRNA | BC103010.1 | - | 1 - 2 | 123 | 82 | |
DV802141.1 | - | 1 - 2 | 116 | 82 | ||
EH165860.1 | - | 1 - 2 | 128 | 82 | ||
X53268.1 | - | 69 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |