Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040488.2 | NP_001035578.1 | 1 - 2 | 197 | 129 | |
GenBank mRNA | BC102128.1 | - | 1 - 2 | 199 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |