Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040523.2 | NP_001035613.1 | 2 - 3 | 253 | 93 | |
GenBank mRNA | BC109742.1 | - | 2 - 3 | 254 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |