Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040594.2 | NP_001035684.1 | 4 - 5 | 508 | 62 | |
GenBank mRNA | BC103147.1 | - | 4 - 5 | 509 | 62 | |
BC126491.1 | - | 5 - 6 | 1951 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |