Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040595.3 | NP_001035685.2 | 3 - 4 | 407 | 56 | |
GenBank mRNA | BC105216.1 | - | 2 - 3 | 216 | 56 | |
BC149602.1 | - | 3 - 4 | 372 | 56 | ||
EW684597.1 | - | 3 - 4 | 407 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |