Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075483.2 | NP_001068951.1 | 1 - 2 | 56 | 70 | |
GenBank mRNA | BC103011.1 | - | 1 - 2 | 57 | 70 | |
BT030535.1 | - | 2 - 3 | 102 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |