Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076490.2 | NP_001069958.1 | 2 - 3 | 239 | 108 | |
GenBank mRNA | BC102201.1 | - | 2 - 3 | 240 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |