Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076514.2 | NP_001069982.1 | 3 - 4 | 255 | 91 | |
GenBank mRNA | BC109734.1 | - | 3 - 4 | 256 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |