Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077045.2 | NP_001070513.1 | 1 - 2 | 122 | 58 | |
GenBank mRNA | BC103446.1 | - | 1 - 2 | 123 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |