Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077046.1 | NP_001070514.1 | 2 - 3 | 153 | 83 | |
GenBank mRNA | BC104531.1 | - | 2 - 3 | 153 | 83 | |
BT025413.1 | - | 2 - 3 | 131 | 83 | ||
BT026183.1 | - | 2 - 3 | 135 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |