Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077046.1 | NP_001070514.1 | 7 - 8 | 1053 | 61 | |
GenBank mRNA | BC104531.1 | - | 7 - 8 | 1053 | 61 | |
BT025413.1 | - | 7 - 8 | 1031 | 61 | ||
BT026183.1 | - | 7 - 8 | 1035 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |